2-Bromo-N-methylbenzamide 98%

Supplier: Ambeed
Warning

A803026-1G A803026-250MG
77268-258EA 22 USD
77268-258 77268-260
2-Bromo-N-methylbenzamide 98%
2-Bromo-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD00099250
CAS Number: 61436-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR