2-Ethylthiophene 98%

Supplier: Ambeed
Danger

Synonyms: o-Ethylthiophene

A801806-25G A801806-5G
77268-246EA 62.83 USD
77268-246 77268-248
2-Ethylthiophene 98%
2-Ethylthiophene

Precaución: for Research Use Only

Formula: C₆H₈S
MW: 112.2 g/mol
MDL Number: MFCD00005461
CAS Number: 872-55-9
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR