6-Bromo-7-methylpyrazolo[1,5-a]pyridine 97%

Supplier: Ambeed
Warning

A801618-100MG A801618-250MG
77268-242EA 101.97 USD
77268-242 77268-244
6-Bromo-7-methylpyrazolo[1,5-a]pyridine 97%
6-Bromo-7-methylpyrazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.05 g/mol
MDL Number: MFCD23710616
CAS Number: 1427382-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR