2-(2,4,5-Trifluorophenyl)ethanol 98%
Supplier: Ambeed
|
Warning
|
A801116-1G
A801116-250MG
A801116-5G
77244-216EA
23
USD
77244-216
77244-218
77244-220
2-(2,4,5-Trifluorophenyl)ethanol 98%
2-(2,4,5-Trifluorophenyl)ethanol
Precaución: for Research Use Only
Formula:
C₈H₇F₃O |
MDL Number:
MFCD06660356 CAS Number: 883267-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...