(S)-2-Methoxypropan-1-amine 98%
Supplier: Ambeed
|
Danger
|
A801095-250MG
A801095-1G
77255-974EA
69
USD
77255-974
77552-464
(S)-2-Methoxypropan-1-amine 98%
(S)-2-Methoxypropan-1-amine
Precaución: for Research Use Only
Formula:
C₄H₁₁NO |
MDL Number:
MFCD06213211 CAS Number: 907943-71-9 UN: 2733 ADR: 3,8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...