2-Chloro-4-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A800923-1G A800923-25G A800923-5G
77268-236EA 41.2 USD
77268-236 77268-238 77268-240
2-Chloro-4-iodobenzoic acid 98%
2-Chloro-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClIO₂
MW: 282.46 g/mol
MDL Number: MFCD05863523
CAS Number: 145343-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR