4'-Isopropylacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: p-Isopropylacetophenone, 4-Isopropylacetophenone

A800596-5G A800596-25G
77293-794EA 7.21 USD
77293-794 77573-560
4'-Isopropylacetophenone 98%
4'-Isopropylacetophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O
MW: 162.23 g/mol
MDL Number: MFCD00048297
CAS Number: 645-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR