2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone 95%

Supplier: Ambeed
Warning

A800433-100G A800433-25G A800433-5G
77255-968EA 51 USD
77255-968 77255-970 77255-972
2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone 95%
2,5-Bis(1,1,3,3-tetramethylbutyl)hydroquinone

Precaución: for Research Use Only

Formula: C₂₂H₃₈O₂
MDL Number: MFCD00128797
CAS Number: 903-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR