1-Ethynyl-1-cyclohexene 97%

Supplier: Ambeed
Danger

Synonyms: (1-Cyclohexenyl)acetylene, 1-Ethynylcyclohexene

A799946-1G A799946-5G
77244-214EA 11.33 USD
77244-214 77552-460
1-Ethynyl-1-cyclohexene 97%
1-Ethynyl-1-cyclohexene

Precaución: for Research Use Only

Formula: C₈H₁₀
MW: 106.17 g/mol
MDL Number: MFCD00001568
CAS Number: 931-49-7
UN: 3295
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR