2,2-Diphenylpropane 98%

Supplier: Ambeed
Warning

Synonyms: Propane-2,2-diyldibenzene

A799554-1G A799554-250MG A799554-25G A799554-5G
77255-960EA 73 USD
77255-960 77255-962 77255-964 77255-966
2,2-Diphenylpropane 98%
2,2-Diphenylpropane

Precaución: for Research Use Only

Formula: (C₆H₅)₂C(CH₃)₂
MDL Number: MFCD00008794
CAS Number: 778-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR