2-Iodo-3,5-dimethylpyrazine 97%

Supplier: Ambeed
Warning

A799102-1G A799102-250MG
77268-226EA 180.25 USD
77268-226 77268-228
2-Iodo-3,5-dimethylpyrazine 97%
2-Iodo-3,5-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MW: 234.03 g/mol
MDL Number: MFCD25969842
CAS Number: 99969-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR