1-Amino-2-methoxyethane 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methoxyethylamine

A799079-25G A799079-5G A799079-10G
77268-222EA 15.45 USD
77268-222 77268-224 77552-454
1-Amino-2-methoxyethane 98%
1-Amino-2-methoxyethane

Precaución: for Research Use Only

Formula: C₃H₉NO
MW: 75.11 g/mol
MDL Number: MFCD00008180
CAS Number: 109-85-3
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR