3-Iodo-5-methylpyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A797110-1G A797110-250MG A797110-5G
77268-218EA 51.5 USD
77268-218 77268-220 77749-572
3-Iodo-5-methylpyridin-2(1H)-one 98%
3-Iodo-5-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆INO
MW: 235.02 g/mol
MDL Number: MFCD27923660
CAS Number: 313678-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR