2-(Ethylthio)Ethylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Ethylthio)ethanamine hydrochloride, 2-(Ethylthio)ethanamine hydrochloride(Flake or chunks or pellets)

A796965-100MG A796965-1G A796965-250MG
77255-950EA 31.93 USD
77255-950 77255-952 77255-954
2-(Ethylthio)Ethylamine hydrochloride 98%
2-(Ethylthio)Ethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁NS·ClH
MW: 141.663 g/mol
MDL Number: MFCD00012903
CAS Number: 54303-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR