3-Ethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A796945-100G A796945-25G A796945-5G
77255-944EA 246 USD
77255-944 77255-946 77255-948
3-Ethoxybenzaldehyde 98%
3-Ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MDL Number: MFCD00016606
CAS Number: 22924-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR