(S)-1-Amino-8-(diphenylphosphino)-1,2,3,4-tetrahydronaphthalene 98%

Supplier: Ambeed
Warning

A796548-1G A796548-250MG A796548-50MG A796548-100MG
77255-932EA 319.3 USD
77255-932 77255-934 77255-936 77747-547
(S)-1-Amino-8-(diphenylphosphino)-1,2,3,4-tetrahydronaphthalene 98%
(S)-1-Amino-8-(diphenylphosphino)-1,2,3,4-tetrahydronaphthalene

Precaución: for Research Use Only

Formula: C₂₂H₂₂NP
MW: 331.39 g/mol
MDL Number: MFCD17013991
CAS Number: 1222630-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR