3-(Benzo[d]thiazol-2-yl)-6-ethyl-7-hydroxy-8-(piperidin-1-ylmethyl)-4H-chromen-4-one 99%

Supplier: Ambeed
Warning

A796427-10MG A796427-1MG A796427-25MG A796427-50MG A796427-5MG A796427-100MG A796427-250MG
77244-188EA 80 USD
77244-188 77244-190 77244-192 77244-194 77244-196 77618-214 77747-546
3-(Benzo[d]thiazol-2-yl)-6-ethyl-7-hydroxy-8-(piperidin-1-ylmethyl)-4H-chromen-4-one 99%
3-(Benzo[d]thiazol-2-yl)-6-ethyl-7-hydroxy-8-(piperidin-1-ylmethyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₂O₃S
MDL Number: MFCD00711481
CAS Number: 222716-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR