(11bS)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S,R,R)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine, (+)-N,N-Bis[(1R)-1-phenylethyl]-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine, (11bS)
A796324-100MG
A796324-1G
A796324-250MG
A796324-25G
A796324-5G
77255-922EA
41.2
USD
77255-922
77255-924
77255-926
77255-928
77255-930
(11bS)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥98%, ee 99%
(11bS)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine
Precaución: for Research Use Only
Formula:
C₃₆H₃₀NO₂P MW: 539.60 g/mol |
MDL Number:
MFCD08561138 CAS Number: 497883-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...