α-Bromo-2,3,5,6-tetrafluorotoluene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,3,5,6-Tetrafluorobenzyl bromide, 2,3,5,6-Tetrafluorobenzylbromide
A796257-1G
A796257-250MG
A796257-25G
A796257-5G
77255-914EA
24.72
USD
77255-914
77255-916
77255-918
77255-920
α-Bromo-2,3,5,6-tetrafluorotoluene 98%
α-Bromo-2,3,5,6-tetrafluorotoluene
Precaución: for Research Use Only
Formula:
C₇H₃BrF₄ MW: 243 g/mol |
MDL Number:
MFCD00013549 CAS Number: 53001-73-3 UN: 3265 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...