3-[4-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole 97%

Supplier: Ambeed
Danger

A795995-1G A795995-5G
77281-388EA 27 USD
77281-388 77281-390
3-[4-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole 97%
3-[4-(Chloromethyl)phenyl]-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂
MDL Number: MFCD09702351
CAS Number: 916766-83-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR