6-(2,6-Dichlorophenyl)-2-((4-(2-(diethylamino)ethoxy)phenyl)amino)-8-methylpyrido[2,3-d]pyrimidin-7(8H)-one 98%
Supplier: Ambeed
|
Warning
|
A795383-10MG
A795383-1MG
A795383-25MG
A795383-5MG
77268-210EA
38
USD
77268-210
77268-212
77268-214
77268-216
6-(2,6-Dichlorophenyl)-2-((4-(2-(diethylamino)ethoxy)phenyl)amino)-8-methylpyrido[2,3-d]pyrimidin-7(8H)-one 98%
6-(2,6-Dichlorophenyl)-2-((4-(2-(diethylamino)ethoxy)phenyl)amino)-8-methylpyrido[2,3-d]pyrimidin-7(8H)-one
Precaución: for Research Use Only
Formula:
C₂₆H₂₇Cl₂N₅O₂ |
MDL Number:
MFCD00950060 CAS Number: 185039-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...