4-(2-Fluoro-4-nitrophenyl)morpholine 98%
Supplier: Ambeed
|
Warning
|
A794986-100G
A794986-25G
A794986-500G
A794986-5G
77293-762EA
113.3
USD
77293-762
77293-764
77293-766
77293-768
4-(2-Fluoro-4-nitrophenyl)morpholine 98%
4-(2-Fluoro-4-nitrophenyl)morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₁₁FN₂O₃ MW: 226.20 g/mol |
MDL Number:
MFCD03138381 CAS Number: 2689-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...