2,3-Dimercapto-1-propanol 99%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dimercapto-2,3 propanol-1, Dithioglycerine, 2,3-Dimercaptopropanol, BAL, Dimercaprol, 2,3-Dimercaptopropan-1-ol, 2,3-Dimercaptopropane-1-ol
A794824-1G
A794824-250MG
A794824-5G
77293-756EA
33.99
USD
77293-756
77293-758
77293-760
2,3-Dimercapto-1-propanol 99%
2,3-Dimercapto-1-propanol
Precaución: for Research Use Only
Formula:
C₃H₈OS₂ MW: 124.22 g/mol |
MDL Number:
MFCD00004864 CAS Number: 59-52-9 UN: 2810 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...