3-Hydroxyoctadecanoic acid 97%

Supplier: Ambeed
Warning

A794298-100MG A794298-250MG
77244-182EA 121 USD
77244-182 77244-184
3-Hydroxyoctadecanoic acid 97%
3-Hydroxyoctadecanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₃
MDL Number: MFCD00673437
CAS Number: 17773-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR