1-Formyl-4-piperidinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Formylpiperidine-4-carboxylic acid, 1-Formylisonipecotic Acid

A794291-100G A794291-1G A794291-25G A794291-5G
77244-174EA 161.71 USD
77244-174 77244-176 77244-178 77244-180
1-Formyl-4-piperidinecarboxylic acid 98%
1-Formyl-4-piperidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.17 g/mol
MDL Number: MFCD03425206
CAS Number: 84163-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR