3-(2-Thienyl)pyridine 98%

Supplier: Ambeed
Warning

A794117-100MG A794117-1G A794117-250MG A794117-5G
77268-190EA 16 USD
77268-190 77268-192 77268-194 77268-196
3-(2-Thienyl)pyridine 98%
3-(2-Thienyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₇NS
MDL Number: MFCD01860052
CAS Number: 21298-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR