2,3,4,5-Tetrafluorobenzonitrile 95%

Supplier: Ambeed
Danger

A793118-1G A793118-25G A793118-5G
77255-898EA 22.66 USD
77255-898 77255-900 77255-902
2,3,4,5-Tetrafluorobenzonitrile 95%
2,3,4,5-Tetrafluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇HF₄N
MW: 175.09 g/mol
MDL Number: MFCD00040187
CAS Number: 16582-93-7
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR