3,5-Di-tert-butyl-4-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A793056-100G A793056-5G A793056-25G A793056-500G
77281-376EA 46.35 USD
77281-376 77573-536 77749-564 77749-565
3,5-Di-tert-butyl-4-hydroxybenzoic acid 98%
3,5-Di-tert-butyl-4-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
MDL Number: MFCD00008827
CAS Number: 1421-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR