(15aS)-1,15-Bis(diphenylphosphino)-7,8,9,10-tetrahydro-6H-dibenzo[b,d][1,6]dioxacycloundecine 98%

Supplier: Ambeed
Warning

A792397-100MG A792397-1G
77268-186EA 127 USD
77268-186 77268-188
(15aS)-1,15-Bis(diphenylphosphino)-7,8,9,10-tetrahydro-6H-dibenzo[b,d][1,6]dioxacycloundecine 98%
(15aS)-1,15-Bis(diphenylphosphino)-7,8,9,10-tetrahydro-6H-dibenzo[b,d][1,6]dioxacycloundecine

Precaución: for Research Use Only

Formula: C₄₁H₃₆O₂P₂
CAS Number: 486429-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR