Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II) 98%

Supplier: Ambeed
Warning

Synonyms: Dichlorobis[2-(di-t-butylphosphino)ethylamine]ruthenium(II), DICHLOROBIS[2-(DI-T-BUTYLPHOSPHINO)ETHYLAMINE]RUTHENIUM(II)

A791783-100MG A791783-1G A791783-250MG
77255-892EA 32 USD
77255-892 77255-894 77255-896
Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II) 98%
Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)

Precaución: for Research Use Only

Formula: C₂₀H₄₈Cl₂N₂P₂Ru
MDL Number: MFCD07782003
CAS Number: 1092372-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR