2-(1H-Pyrrol-1-yl)thiophene-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A791617-100MG A791617-1G A791617-250MG A791617-5G
77244-148EA 55.62 USD
77244-148 77244-150 77244-152 77244-154
2-(1H-Pyrrol-1-yl)thiophene-3-carboxylic acid 97%
2-(1H-Pyrrol-1-yl)thiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂S
MDL Number: MFCD01871358
CAS Number: 79242-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR