(S)-2-Amino-6-(3-(3-(3-methyl-3H-diazirin-3-yl)propyl)ureido)hexanoic acid 98%

Supplier: Ambeed
Danger

Synonyms: DiZPK

A791385-10MG A791385-1MG A791385-100MG
77244-142EA 94.76 USD
77244-142 77244-144 77747-540
(S)-2-Amino-6-(3-(3-(3-methyl-3H-diazirin-3-yl)propyl)ureido)hexanoic acid 98%
(S)-2-Amino-6-(3-(3-(3-methyl-3H-diazirin-3-yl)propyl)ureido)hexanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₃N₅O₃
MW: 285.34 g/mol
MDL Number: MFCD28900721
CAS Number: 1337883-32-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR