3-Chloro-4-cyanobenzoic acid 97%

Supplier: Ambeed
Warning

A790819-100MG A790819-1G A790819-250MG
77255-868EA 72.1 USD
77255-868 77255-870 77255-872
3-Chloro-4-cyanobenzoic acid 97%
3-Chloro-4-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂
MW: 181.57 g/mol
MDL Number: MFCD18392433
CAS Number: 1261685-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR