4-Ethynyltriphenylamine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Ethynyl-N,N-diphenylaniline, 4-Diphenylaminophenylacetylene

A790699-1G A790699-250MG A790699-25G A790699-5G
77244-126EA 37 USD
77244-126 77244-128 77244-130 77244-132
4-Ethynyltriphenylamine 97%
4-Ethynyltriphenylamine

Precaución: for Research Use Only

Formula: C₂₀H₁₅N
MDL Number: MFCD09030769
CAS Number: 205877-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR