1,2-Di(pyridin-2-yl)ethane-1,2-dione 98%

Supplier: Ambeed
Warning

A790697-1G A790697-25G A790697-5G
77255-862EA 13.39 USD
77255-862 77255-864 77255-866
1,2-Di(pyridin-2-yl)ethane-1,2-dione 98%
1,2-Di(pyridin-2-yl)ethane-1,2-dione

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₂
MW: 212.20 g/mol
MDL Number: MFCD00006301
CAS Number: 492-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR