3-Acetylbenzonitrile 98%

Supplier: Ambeed
Warning

A790566-100G A790566-10G A790566-1G A790566-25G A790566-5G
77268-162EA 139 USD
77268-162 77268-164 77268-166 77268-168 77268-170
3-Acetylbenzonitrile 98%
3-Acetylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₇NO
MDL Number: MFCD00001806
CAS Number: 6136-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR