2-(3-Benzoylphenyl)propionitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Benzoylphenyl)propanenitrile, Ketoprofen Related Compound F, 3-Benzoyl-alpha-methylphenylacetonitrile, 3-(1-Cyanoethyl)benzophenone

A790557-1G A790557-250MG
77281-370EA 34 USD
77281-370 77281-372
2-(3-Benzoylphenyl)propionitrile 98%
2-(3-Benzoylphenyl)propionitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO
MW: 235.29 g/mol
MDL Number: MFCD00044435
CAS Number: 42872-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR