3-(2,4-Dimethylphenyl)acrylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dimethylcinnamic acid

A790345-1G A790345-250MG A790345-5G
77255-854EA 27.81 USD
77255-854 77255-856 77255-858
3-(2,4-Dimethylphenyl)acrylic acid 95%
3-(2,4-Dimethylphenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD00017287
CAS Number: 1685-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR