2-Fluoro-5-(methylsulfonamido)benzoic acid 97%

Supplier: Ambeed
Warning

A790325-1G A790325-250MG
77268-154EA 70.04 USD
77268-154 77268-156
2-Fluoro-5-(methylsulfonamido)benzoic acid 97%
2-Fluoro-5-(methylsulfonamido)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈FNO₄S
MW: 233.22 g/mol
MDL Number: MFCD12149130
CAS Number: 137315-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR