TG003 99+%

Supplier: Ambeed
Warning

A789987-100MG A789987-10MG A789987-1MG A789987-25MG A789987-50MG A789987-5MG
77268-142EA 122 USD
77268-142 77268-144 77268-146 77268-148 77268-150 77268-152
TG003 99+%
(E/Z)-TG003

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₂S
MDL Number: MFCD00624584
CAS Number: 300801-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR