1-Bromo-3-(trifluoromethylthio)benzene 97%

Supplier: Ambeed
Danger

Synonyms: (3-Bromophenyl)(trifluoromethyl)sulphane

A789865-100MG A789865-10G A789865-1G A789865-250MG A789865-25G A789865-5G
77281-358EA 14.42 USD
77281-358 77281-360 77281-362 77281-364 77281-366 77281-368
1-Bromo-3-(trifluoromethylthio)benzene 97%
1-Bromo-3-(trifluoromethylthio)benzene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃S
MW: 257.0709 g/mol
MDL Number: MFCD00236348
CAS Number: 2252-45-1
UN: 1760
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR