Methyl 4-nitro-1H-indazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A788993-1G A788993-250MG A788993-5G
77293-742EA 14 USD
77293-742 77293-744 77293-746
Methyl 4-nitro-1H-indazole-6-carboxylate 97%
Methyl 4-nitro-1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇N₃O₄
MDL Number: MFCD07368219
CAS Number: 72922-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR