4,4-Dimethyloxazolidine 95%

Supplier: Ambeed
Danger

A788847-100MG A788847-1G A788847-250MG A788847-5G
77293-734EA 130.81 USD
77293-734 77293-736 77293-738 77293-740
4,4-Dimethyloxazolidine 95%
4,4-Dimethyloxazolidine

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.14 g/mol
MDL Number: MFCD00154171
CAS Number: 51200-87-4
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR