4-(2-Oxopropyl)benzonitrile 96%

Supplier: Ambeed
Warning

A788767-100MG A788767-1G
77293-730EA 42 USD
77293-730 77293-732
4-(2-Oxopropyl)benzonitrile 96%
4-(2-Oxopropyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD02094171
CAS Number: 58949-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR