Sodium-4-aminonaphthalene-1-sulphonate tetrahydrate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Naphthylamine-4-sulphonic acid sodium salt tetrahydrate, 4-Aminonaphthalene-1-sulphonic acid sodium salt tetrahydrate, Sodium 4-Amino-1-naphthalenesulphonate tetrahydrate, Sodium naphthionate tetrahydrate, 4-Amino-α-naphthalenesulphonic acid sodium salt tetrahydrate, 4-Amino-1-naphthalenesulphonic acid sodium salt tetrahydrate, Naphthionic acid sodium salt tetrahydrate, Sodium 1-naphthylamine-4-sulphonate tetrahydrate

A788153-1000G A788153-100G A788153-10G A788153-25G A788153-500G
77293-712EA 128 USD
77293-712 77293-714 77293-716 77293-718 77293-720
Sodium-4-aminonaphthalene-1-sulphonate tetrahydrate 95%
Sodium-4-aminonaphthalene-1-sulphonate tetrahydrate

Precaución: for Research Use Only

Formula: C₁₀H₈NNaO₃S.4H₂O
MW: 245.23 g/mol
Melting Pt: 280
MDL Number: MFCD00044844
CAS Number: 130-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR