3-Chloro-4-hydroxyphenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
A787735-1G
A787735-250MG
A787735-5G
77281-338EA
70.04
USD
77281-338
77281-340
77281-342
3-Chloro-4-hydroxyphenylboronic acid pinacol ester 98%
3-Chloro-4-hydroxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BClO₃ MW: 254.5200 g/mol |
MDL Number:
MFCD09260490 CAS Number: 629658-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...