(±)-3-Heptanol 98%

Supplier: Ambeed
Danger

A786532-25G A786532-5G
77281-322EA 52 USD
77281-322 77281-324
(±)-3-Heptanol 98%
(±)-3-Heptanol

Precaución: for Research Use Only

Formula: C₇H₁₆O
MDL Number: MFCD00004586
CAS Number: 589-82-2
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR