2,3,6-Trifluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A786401-1G A786401-25G A786401-5G
77293-698EA 14.42 USD
77293-698 77293-700 77293-702
2,3,6-Trifluorophenylboronic acid 98%
2,3,6-Trifluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
MDL Number: MFCD01863167
CAS Number: 247564-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR