4-(Bromomethyl)-2-fluoropyridine 97%

Supplier: Ambeed
Danger

A785702-100MG A785702-250MG
77268-100EA 212 USD
77268-100 77268-102
4-(Bromomethyl)-2-fluoropyridine 97%
4-(Bromomethyl)-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MDL Number: MFCD09953531
CAS Number: 64992-03-6
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR