2-(Methylthio)ethanol 95%

Supplier: Ambeed
Warning

A785499-100G A785499-25G
77255-834EA 69 USD
77255-834 77255-836 77255-838
2-(Methylthio)ethanol 95%
2-(Methylthio)ethanol

Precaución: for Research Use Only

Formula: C₃H₈OS
MDL Number: MFCD00002908
CAS Number: 5271-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR